4-(Trifluoromethyl)cyclohexanone 97%
Supplier: Apollo Scientific
|
Warning
|
PC7668N-1G
PC7668N-5G
PC7668N-100MG
APOSPC7668N-1GEA
94.4
EUR
APOSPC7668N-1G
APOSPC7668N-5G
APOSPC7668N-100MG
4-(Trifluoromethyl)cyclohexanone 97%
4-(Trifluoromethyl)cyclohexanone
Formula:
C₇H₉F₃O Boiling Pt: 67…68 °C/16mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00102145 CAS Number: 75091-99-5 UN: 1224 ADR: 3,III |