3-(Trifluoromethyl)-3-hydroxybutyric acid 95%
Supplier: Apollo Scientific
Danger
PC7671-1G PC7671-25G PC7671-5G
APOSPC7671-1GEA 51 EUR
APOSPC7671-1G APOSPC7671-25G APOSPC7671-5G
3-(Trifluoromethyl)-3-hydroxybutyric acid 95%
3-(Trifluoromethyl)-3-hydroxybutyric acid
Formula: C₅H₇F₃O₃
Boiling Pt: 257 °C
Melting Pt: 39…41 °C
Storage Temperature: Ambient
MDL Number: MFCD00039531
CAS Number: 338-03-4
UN: 3261
ADR: 8,III