5-(Trifluoromethyl)pyridin-2-ylamidoxime 95+%
Supplier: Apollo Scientific
Warning
Synonyms: 5-(Trifluoromethyl)pyridine-2-carboxamide oxime
PC7685-1G PC7685-5G
APOSPC7685-1GEA 219 EUR
APOSPC7685-1G APOSPC7685-5G
5-(Trifluoromethyl)pyridin-2-ylamidoxime 95+%
5-(Trifluoromethyl)pyridin-2-ylamidoxime
Formula: C₇H₆F₃N₃O
MW: 205.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00205809
CAS Number: 175277-44-8