(3-(Trifluoromethyl)phenoxy)acetonitrile 97%
Supplier: Apollo Scientific
Warning
PC7696-1G PC7696-5G PC7696-10G
APOSPC7696-1GEA 21.9 EUR
APOSPC7696-1G APOSPC7696-5G APOSPC7696-10G
(3-(Trifluoromethyl)phenoxy)acetonitrile 97%
(3-(Trifluoromethyl)phenoxy)acetonitrile
Formula: C₉H₆F₃NO
MW: 201.15 g/mol
Boiling Pt: 82…84 °C/0.5mmHg
Storage Temperature: Ambient
CAS Number: 2145-31-5
UN: 3276
ADR: 6.1,III