2-(Trifluoromethyl)phenylhydrazine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(2-(Trifluoromethyl)phenyl)hydrazine, 2-Hydrazinobenzotrifluoride
PC7716-1G PC7716-5G
APOSPC7716-1GEA 30.6 EUR
APOSPC7716-1G APOSPC7716-5G
2-(Trifluoromethyl)phenylhydrazine 98%
2-(Trifluoromethyl)phenylhydrazine
Formula: C₇H₇F₃N₂
MW: 176.14 g/mol
Melting Pt: 56…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00052687
CAS Number: 365-34-4