3-(Trifluoromethyl)phenylhydrazine 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(3-(Trifluoromethyl)phenyl)hydrazine, 3-Hydrazinobenzotrifluoride
PC7716M-25G PC7716M-5G
APOSPC7716M-25GEA 545 EUR
APOSPC7716M-25G APOSPC7716M-5G
3-(Trifluoromethyl)phenylhydrazine 97%
3-(Trifluoromethyl)phenylhydrazine
Formula: C₇H₇F₃N₂
MW: 176.14 g/mol
Boiling Pt: 140 °C/30mmHg
Density: 1.348 g/cm³ (25 °C)
Flash Pt: 107 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00025093
CAS Number: 368-78-5
EINECS: 206-713-5