2-(Trifluoromethyl)acrylic acid 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2-(Trifluoromethyl)propenoic acid
PC7761-25G PC7761-5G PC7761-100G
APOSPC7761-25GEA 111 EUR
APOSPC7761-25G APOSPC7761-5G APOSPC7761-100G
2-(Trifluoromethyl)acrylic acid 98%
2-(Trifluoromethyl)acrylic acid
Formula: C₄H₃F₃O₂
MW: 140.06 g/mol
Boiling Pt: 86 °C/35mmHg
Melting Pt: 48…52 °C
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00042424
CAS Number: 381-98-6
UN: 3261
ADR: 8,II