2'-(Trifluoromethyl)propiophenone 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(2-(Trifluoromethyl)phenyl)propan-1-one, o-(Trifluoromethyl)propiophenone
PC7764-25G PC7764-5G
APOSPC7764-25GEA 114 EUR
APOSPC7764-25G APOSPC7764-5G
2'-(Trifluoromethyl)propiophenone 97%
2'-(Trifluoromethyl)propiophenone
Formula: C₁₀H₉F₃O
MW: 202.18 g/mol
Boiling Pt: 219…220 °C
Density: 1.214 g/cm³ (25 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00013557
CAS Number: 16185-96-9