3'-(Trifluoromethyl)propiophenone 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(3-(Trifluoromethyl)phenyl)propan-1-one, m-(Trifluoromethyl)propiophenone
PC7765-100G PC7765-25G PC7765-5G
APOSPC7765-100GEA 122 EUR
APOSPC7765-100G APOSPC7765-25G APOSPC7765-5G
3'-(Trifluoromethyl)propiophenone 97%
3'-(Trifluoromethyl)propiophenone
Formula: C₁₀H₉F₃O
Boiling Pt: 216…217 °C
Melting Pt: 18…19 °C
Density: 1.202 g/cm³ (25 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00039227
CAS Number: 1533-03-5