4'-(Trifluoromethyl)propiophenone 97%
Supplier: Apollo Scientific
Warning
Synonyms: p-(Trifluoromethyl)propiophenone, 1-(4-(Trifluoromethyl)phenyl)propan-1-one
PC7766-100G PC7766-25G PC7766-5G
APOSPC7766-100GEA 366 EUR
APOSPC7766-100G APOSPC7766-25G APOSPC7766-5G
4'-(Trifluoromethyl)propiophenone 97%
4'-(Trifluoromethyl)propiophenone
Formula: C₁₀H₉F₃O
MW: 202.18 g/mol
Boiling Pt: 216 °C
Melting Pt: 36…39 °C
Density: 1.973 g/cm³ (25 °C)
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00039231
CAS Number: 711-33-1