3-(Trifluoromethyl)pyridine 97%
Supplier: Apollo Scientific
Danger
PC7766G-1G PC7766G-25G PC7766G-5G
APOSPC7766G-1GEA 21.9 EUR
APOSPC7766G-1G APOSPC7766G-25G APOSPC7766G-5G
3-(Trifluoromethyl)pyridine 97%
3-(Trifluoromethyl)pyridine
Formula: C₆H₄F₃N
MW: 147.1 g/mol
Boiling Pt: 113…115 °C
Density: 1.276 g/cm³ (25 °C)
Flash Pt: 21 °C
Storage Temperature: Ambient
MDL Number: MFCD00100032
CAS Number: 3796-23-4
EINECS: 223-268-2
UN: 1992
ADR: 3+6.1 III