4-(Trifluoromethyl)pyridine
Supplier: Apollo Scientific
Danger
PC7766H-1G PC7766H-5G
APOSPC7766H-1GEA 21.9 EUR
APOSPC7766H-1G APOSPC7766H-5G
4-(Trifluoromethyl)pyridine
4-(Trifluoromethyl)pyridine
Formula: C₆H₄F₃N
MW: 147.1 g/mol
Boiling Pt: 108 °C
Density: 1.27 g/cm³ (25 °C)
Flash Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00153289
CAS Number: 3796-24-5
UN: 1993
ADR: 3 II