4-(Trifluoromethylthio)benzyl bromide 97%
Supplier: Apollo Scientific
Danger
Synonyms: (4-(Bromomethyl)phenyl)(trifluoromethyl)sulphane
PC7769K-1G PC7769K-100MG PC7769K-250MG
APOSPC7769K-1GEA 43.5 EUR
APOSPC7769K-1G APOSPC7769K-100MG APOSPC7769K-250MG
4-(Trifluoromethylthio)benzyl bromide 97%
4-(Trifluoromethylthio)benzyl bromide
Formula: C₈H₆BrF₃S
MW: 271.10 g/mol
Boiling Pt: 115…118 °C/13mmHg
Melting Pt: 53…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00129189
CAS Number: 21101-63-3
UN: 3261
ADR: 8 II