3-(Trifluoromethyl)thiophenol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Mercaptobenzotrifluoride, 3-(Trifluoromethyl)benzenethiol, 3-(Trifluoromethyl)benzene-1-thiol
PC7772-25G PC7772-5G
APOSPC7772-25GEA 272 EUR
APOSPC7772-25G APOSPC7772-5G
3-(Trifluoromethyl)thiophenol 98%
3-(Trifluoromethyl)thiophenol
Formula: C₇H₅F₃S
MW: 178.18 g/mol
Boiling Pt: 71…72 °C/20mmHg
Flash Pt: 68.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041142
CAS Number: 937-00-8
UN: 2810
ADR: 6.1 III