4-(Trifluoromethyl)thiophenol 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Mercaptobenzotrifluoride, 4-(Trifluoromethyl)benzene-1-thiol, 4-(Trifluoromethyl)benzenethiol
PC7773-25G PC7773-50G PC7773-5G
APOSPC7773-25GEA 412 EUR
APOSPC7773-25G APOSPC7773-50G APOSPC7773-5G
4-(Trifluoromethyl)thiophenol 97%
4-(Trifluoromethyl)thiophenol
Formula: C₇H₅F₃S
MW: 178.18 g/mol
Boiling Pt: 66…68 °C/15mmHg
Density: 1.3 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00042320
CAS Number: 825-83-2