4-(Trifluoromethylthio)phenol 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-[(Trifluoromethyl)sulphanyl]phenol
PC7773A-100G PC7773A-25G PC7773A-5G
APOSPC7773A-100GEA 63.8 EUR
APOSPC7773A-100G APOSPC7773A-25G APOSPC7773A-5G
4-(Trifluoromethylthio)phenol 97%
4-(Trifluoromethylthio)phenol
Formula: C₇H₅F₃OS
MW: 194.18 g/mol
Boiling Pt: 77…78 °C
Melting Pt: 57…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00036035
CAS Number: 461-84-7