3-Methoxy-2,4,5-trifluorobenzoyl chloride
Supplier: Apollo Scientific
Danger
Synonyms: 3-Methoxy-2,4,5-trifluorobenzene-1-carbonyl chloride, 2,4,5-Trifluoro-3-methoxybenzoyl chloride
PC7787-25G PC7787-5G PC7787-100G
APOSPC7787-25GEA 30.6 EUR
APOSPC7787-25G APOSPC7787-5G APOSPC7787-100G
3-Methoxy-2,4,5-trifluorobenzoyl chloride
3-Methoxy-2,4,5-trifluorobenzoyl chloride
Formula: C₈H₄ClF₃O₂
MW: 224.56 g/mol
Boiling Pt: 108…112 °C/20mmHg
Density: 1.472 g/cm³ (25 °C)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD02682012
CAS Number: 112811-66-2
UN: 3265
ADR: 8,II