1,2,3-Trifluoro-4-nitrobenzene 99%
Supplier: Apollo Scientific
Warning
Synonyms: 2,3,4-Trifluoronitrobenzene
PC7789-100G PC7789-250G PC7789-25G PC7789-500G PC7789-1KG
APOSPC7789-100GEA 21.9 EUR
APOSPC7789-100G APOSPC7789-250G APOSPC7789-25G APOSPC7789-500G APOSPC7789-1KG
1,2,3-Trifluoro-4-nitrobenzene 99%
1,2,3-Trifluoro-4-nitrobenzene
Formula: C₆H₂F₃NO₂
MW: 177.08 g/mol
Boiling Pt: 92 °C/20mmHg
Density: 1.541 g/cm³ (25 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00041546
CAS Number: 771-69-7
EINECS: 212-238-4
UN: 2810
ADR: 6.1 III