4-Chloro-1-fluoro-2-nitrobenzene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Chloro-2-fluoronitrobenzene
PC7803-25G PC7803-100G PC7803-5G PC7803-500G
APOSPC7803-25GEA 21.9 EUR
APOSPC7803-25G APOSPC7803-100G APOSPC7803-5G APOSPC7803-500G
4-Chloro-1-fluoro-2-nitrobenzene 97%
4-Chloro-1-fluoro-2-nitrobenzene
Formula: C₆H₃ClFNO₂
MW: 175.55 g/mol
Boiling Pt: 237 °C
Melting Pt: 8 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00069418
CAS Number: 345-18-6