5-Fluoro-o-anisaldehyde 99%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Fluoro-2-anisaldehyde, 5-Fluoro-2-methoxybenzaldehyde
PC7804-100G PC7804-25G PC7804-5G
APOSPC7804-100GEA 219 EUR
APOSPC7804-100G APOSPC7804-25G APOSPC7804-5G
5-Fluoro-o-anisaldehyde 99%
5-Fluoro-o-anisaldehyde
Formula: C₈H₇FO₂
MW: 154.14 g/mol
Boiling Pt: 118 °C/14.25mmHg
Melting Pt: 41…48 °C
Storage Temperature: Ambient
MDL Number: MFCD00143458
CAS Number: 19415-51-1