5,7-Difluoroindole
Supplier: Apollo Scientific
Warning
PC7807-1G PC7807-250MG PC7807-5G
APOSPC7807-1GEA 198 EUR
APOSPC7807-1G APOSPC7807-250MG APOSPC7807-5G
5,7-Difluoroindole
5,7-Difluoroindole
Formula: C₈H₅F₂N
MW: 153.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01075215
CAS Number: 301856-25-7