2,3,4-Trifluorophenol 98%
Supplier: Apollo Scientific
Danger
PC7817-100G PC7817-25G PC7817-5G
APOSPC7817-100GEA 319 EUR
APOSPC7817-100G APOSPC7817-25G APOSPC7817-5G
2,3,4-Trifluorophenol 98%
2,3,4-Trifluorophenol
Formula: C₆H₃F₃O
MW: 148.08 g/mol
Boiling Pt: 68…70 °C/43mmHg
Melting Pt: 30…34 °C
Density: 1.46 g/cm³ (25 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00042426
CAS Number: 2822-41-5
UN: 2925
ADR: 4.1+8,III