3,4,5-Trifluorophenol 95%
Supplier: Apollo Scientific
Danger
PC7818-100G PC7818-25G PC7818-5G
APOSPC7818-100GEA 143 EUR
APOSPC7818-100G APOSPC7818-25G APOSPC7818-5G
3,4,5-Trifluorophenol 95%
3,4,5-Trifluorophenol
Formula: C₆H₃F₃O
MW: 148.08 g/mol
Boiling Pt: 177 °C
Melting Pt: 52…55 °C
Density: 1.629
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00042427
CAS Number: 99627-05-1
EINECS: 436-120-9
UN: 3261
ADR: 8 II