(2,3,5-Trifluorophenyl)acetonitrile 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2,3,5-Trifluorobenzyl cyanide, 2,3,5-Trifluorophenylacetonitrile
PC7818K-1G PC7818K-250MG
APOSPC7818K-1GEA 65.4 EUR
APOSPC7818K-1G APOSPC7818K-250MG
(2,3,5-Trifluorophenyl)acetonitrile 97%
(2,3,5-Trifluorophenyl)acetonitrile
Formula: C₈H₄F₃N
MW: 171.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00083546
CAS Number: 243666-14-0
UN: 3276
ADR: 6.1 III