(2,4,5-Trifluorophenyl)acetonitrile 97%
Supplier: Apollo Scientific
Warning
PC7818M-1G PC7818M-5G PC7818M-25G
APOSPC7818M-1GEA 21.9 EUR
APOSPC7818M-1G APOSPC7818M-5G APOSPC7818M-25G
(2,4,5-Trifluorophenyl)acetonitrile 97%
(2,4,5-Trifluorophenyl)acetonitrile
Formula: C₈H₄F₃N
MW: 171.12 g/mol
Boiling Pt: 84 °C/1mmHg
Density: 1.35 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00061220
CAS Number: 220141-74-2
UN: 3276
ADR: 6.1,III