1-(4-(Trifluoromethyl)phenyl)biguanide hydrochloride
Supplier: Apollo Scientific
Warning
PC7825-25G PC7825-5G
APOSPC7825-25GEA 208 EUR
APOSPC7825-25G APOSPC7825-5G
1-(4-(Trifluoromethyl)phenyl)biguanide hydrochloride
1-(4-(Trifluoromethyl)phenyl)biguanide hydrochloride
Formula: C₉H₁₁ClF₃N₅
Melting Pt: 209…213 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD01569535
CAS Number: 18960-29-7