3-Bromo-4,5-difluoroaniline 98%
Supplier: Apollo Scientific
Warning
PC7859-1G PC7859-5G PC7859-25G
APOSPC7859-1GEA 29 EUR
APOSPC7859-1G APOSPC7859-5G APOSPC7859-25G
3-Bromo-4,5-difluoroaniline 98%
3-Bromo-4,5-difluoroaniline
Formula: C₆H₄BrF₂N
MW: 208.01 g/mol
Melting Pt: 66…69 °C
Storage Temperature: Ambient
MDL Number: MFCD08062370
CAS Number: 875664-41-8
UN: 2811
ADR: 6.1 III