2-Bromo-5-fluoroanisole 98+%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-4-fluoro-2-methoxybenzene
PC7888-25G PC7888-5G PC7888-100G
APOSPC7888-25GEA 31.9 EUR
APOSPC7888-25G APOSPC7888-5G APOSPC7888-100G
2-Bromo-5-fluoroanisole 98+%
2-Bromo-5-fluoroanisole
Formula: C₇H₆BrFO
MW: 205.03 g/mol
Boiling Pt: 143…145 °C
Density: 1.598 g/cm³ (25 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD04973752
CAS Number: 450-88-4