4-Hydroxy-8-methoxy-2-(trifluoromethyl)quinoline 96%
Supplier: Apollo Scientific
Warning
PC7893-1G PC7893-5G PC7893-250MG
APOSPC7893-1GEA 168 EUR
APOSPC7893-1G APOSPC7893-5G APOSPC7893-250MG
4-Hydroxy-8-methoxy-2-(trifluoromethyl)quinoline 96%
4-Hydroxy-8-methoxy-2-(trifluoromethyl)quinoline
Formula: C₁₁H₈F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD01862071
CAS Number: 41192-84-1