1,2-Dibromotrifluoroethyl trifluoromethylether 97%
Supplier: Apollo Scientific
PC7895-25G PC7895-5G
APOSPC7895-25GEA 297 EUR
APOSPC7895-25G APOSPC7895-5G
1,2-Dibromotrifluoroethyl trifluoromethylether 97%
1,2-Dibromotrifluoroethyl trifluoromethylether
Formula: C₃Br₂F₆O
MW: 325.84 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01320784
CAS Number: 2356-57-2