2H-Perfluoro-5-methyl-3,6-dioxanonane 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1,1,1,2,2,3,3-Heptafluoro-3-[[1,1,1,2,3,3-hexafluoro-3-(1,2,2,2-tetrafluoroethoxy)propan-2-yl]oxy]propane
PC7896-25G PC7896-5G PC7896-250MG
APOSPC7896-25GEA 609 EUR
APOSPC7896-25G APOSPC7896-5G APOSPC7896-250MG
2H-Perfluoro-5-methyl-3,6-dioxanonane 97%
2H-Perfluoro-5-methyl-3,6-dioxanonane
Formula: C₈HF₁₇O₂
MW: 452.07 g/mol
Boiling Pt: 103…105 °C
Melting Pt: –122 °C
Density: 1.659 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00054714
CAS Number: 3330-14-1