1,2-Dichloro-3,3,3-trifluoropropene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1,2-Dichloro-3,3,3-trifluoroprop-1-ene
PC7899-1G PC7899-5G
APOSPC7899-1GEA 84.1 EUR
APOSPC7899-1G APOSPC7899-5G
1,2-Dichloro-3,3,3-trifluoropropene 98%
1,2-Dichloro-3,3,3-trifluoropropene
Formula: C₃HCl₂F₃
MW: 164.94 g/mol
Boiling Pt: 53…55 °C
Melting Pt: –109.2 °C
Density: 1.4653 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD08461577
CAS Number: 431-27-6