4-Bromo-2-fluoro-6-(trifluoromethyl)aniline 98%
Supplier: Apollo Scientific
Warning
PC7901-10G PC7901-5G PC7901-1G
APOSPC7901-10GEA 422 EUR
APOSPC7901-10G APOSPC7901-5G APOSPC7901-1G
4-Bromo-2-fluoro-6-(trifluoromethyl)aniline 98%
4-Bromo-2-fluoro-6-(trifluoromethyl)aniline
Formula: C₇H₄BrF₄N
Storage Temperature: Ambient
CAS Number: 875664-46-3