4-Fluoro-2-methoxy-N-methylaniline 96%
Supplier: Apollo Scientific
PC7917-1G PC7917-250MG
APOSPC7917-1GEA 148 EUR
APOSPC7917-1G APOSPC7917-250MG
4-Fluoro-2-methoxy-N-methylaniline 96%
4-Fluoro-2-methoxy-N-methylaniline
Formula: C₈H₁₀FNO
Storage Temperature: Refrigerator
MDL Number: MFCD09475904
CAS Number: 941294-13-9