3-Hydroxy-5-(trifluoromethoxy)phenylboronic acid 98%
Supplier: Apollo Scientific
|
Warning
|
PC7918-1G
PC7918-500MG
APOSPC7918-1GEA
342
EUR
APOSPC7918-1G
APOSPC7918-500MG
3-Hydroxy-5-(trifluoromethoxy)phenylboronic acid 98%
3-Hydroxy-5-(trifluoromethoxy)phenylboronic acid
Formula:
C₇H₆BF₃O₄ Melting Pt: 78…81 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09475898 CAS Number: 957120-22-8 |