2,4,6-Tris(trifluoromethyl)-1,3,5-triazine 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2,4,6-Tris(trifluoromethyl)-s-triazine
PC7920-1G PC7920-25G PC7920-5G
APOSPC7920-1GEA 108 EUR
APOSPC7920-1G APOSPC7920-25G APOSPC7920-5G
2,4,6-Tris(trifluoromethyl)-1,3,5-triazine 97%
2,4,6-Tris(trifluoromethyl)-1,3,5-triazine
Mass spectrometry standard.
Formula: C₆F₉N₃
MW: 285.07 g/mol
Boiling Pt: 98…99 °C/748mmHg
Density: 1.596 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00042436
CAS Number: 368-66-1
EINECS: 206-709-3
UN: 2810
ADR: 6.1 III