1,3,5-Tris[(3,3,3-trifluoropropyl)methyl]cyclotrisiloxane
Supplier: Apollo Scientific
Danger
Synonyms: 1,3,5-Tris(3,3,3-trifluoropropyl)-1,3,5-trimethylcyclotrisiloxane, 1,3,5-Trimethyl-1,3,5-tris(3,3,3-trifluoropropyl)cyclotrisiloxane
PC7925-25G PC7925-100G
APOSPC7925-25GEA 154 EUR
APOSPC7925-25G APOSPC7925-100G
1,3,5-Tris[(3,3,3-trifluoropropyl)methyl]cyclotrisiloxane
1,3,5-Tris[(3,3,3-trifluoropropyl)methyl]cyclotrisiloxane
Formula: C₁₂H₂₁F₉O₃Si₃
MW: 468.54 g/mol
Boiling Pt: 135 °C/22mmHg
Melting Pt: 34…36 °C
Density: 1.24 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00054667
CAS Number: 2374-14-3
EINECS: 219-154-7