4,5-Dichloro-2-fluorobenzenesulphonyl chloride 97%
Supplier: Apollo Scientific
Danger
Synonyms: 4,5-Dichloro-2-fluorobenzene-1-sulphonyl chloride
PC7933-1G PC7933-5G
APOSPC7933-1GEA 87 EUR
APOSPC7933-1G APOSPC7933-5G
4,5-Dichloro-2-fluorobenzenesulphonyl chloride 97%
4,5-Dichloro-2-fluorobenzenesulphonyl chloride
Formula: C₆H₂Cl₃FO₂S
MW: 263.501 g/mol
Melting Pt: 56…57.5 °C
Storage Temperature: Ambient
MDL Number: MFCD03094453
CAS Number: 13656-52-5
UN: 3261
ADR: 8,III