2,3,5,6-Tetrafluorobenzaldehyde
Supplier: Apollo Scientific
Warning
PC7934-25G PC7934-5G
APOSPC7934-25GEA 203 EUR
APOSPC7934-25G APOSPC7934-5G
2,3,5,6-Tetrafluorobenzaldehyde
2,3,5,6-Tetrafluorobenzaldehyde
Formula: C₇H₂F₄O
MW: 178.09 g/mol
Boiling Pt: 178 °C
Density: 1.525 g/cm³ (25 °C)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00167387
CAS Number: 19842-76-3