3-(Trifluoromethyl)-2-pyridinethiol 97%
Supplier: Apollo Scientific
Danger
Synonyms: 3-(Trifluoromethyl)pyridine-2-thiol
PC7961-1G PC7961-5G PC7961-10G
APOSPC7961-1GEA 39.3 EUR
APOSPC7961-1G APOSPC7961-5G APOSPC7961-10G
3-(Trifluoromethyl)-2-pyridinethiol 97%
3-(Trifluoromethyl)-2-pyridinethiol
Formula: C₆H₄F₃NS
Melting Pt: 178…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00178749
CAS Number: 104040-74-6
UN: 2811
ADR: 6.1,III