2-Mercapto-6-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
Warning
Synonyms: 2-Sulphanyl-6-(trifluoromethyl)pyridine
PC7962-1G PC7962-250MG PC7962-5G
APOSPC7962-1GEA 102 EUR
APOSPC7962-1G APOSPC7962-250MG APOSPC7962-5G
2-Mercapto-6-(trifluoromethyl)pyridine
2-Mercapto-6-(trifluoromethyl)pyridine
Formula: C₆H₄F₃NS
MW: 179.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09864680
CAS Number: 121307-80-0