2,3-Difluoro-4-methoxybenzaldehyde 98%
Supplier: Apollo Scientific
Warning
PC7965-5G PC7965-1G PC7965-25G
APOSPC7965-5GEA 156 EUR
APOSPC7965-5G APOSPC7965-1G APOSPC7965-25G
2,3-Difluoro-4-methoxybenzaldehyde 98%
2,3-Difluoro-4-methoxybenzaldehyde
Formula: C₈H₆F₂O₂
Melting Pt: 65…66 °C
Storage Temperature: Ambient
MDL Number: MFCD04115981
CAS Number: 256417-11-5