2,6-Difluoro-3-iodopyridine 97%
Supplier: Apollo Scientific
Warning
PC7983-5G PC7983-1G PC7983-10G
APOSPC7983-5GEA 53.8 EUR
APOSPC7983-5G APOSPC7983-1G APOSPC7983-10G
2,6-Difluoro-3-iodopyridine 97%
2,6-Difluoro-3-iodopyridine
Formula: C₅H₂F₂IN
MW: 240.98 g/mol
Boiling Pt: 85…87 °C/15mmHg
Melting Pt: 41…42 °C
Storage Temperature: Ambient
MDL Number: MFCD09743595
CAS Number: 685517-67-3