2,2,3,3,4,4,4-Heptafluorobutyltrifluoromethanesulphonate 97%
Supplier: Apollo Scientific
|
Danger
|
PC7987-1G
PC7987-5G
APOSPC7987-1GEA
61.3
EUR
APOSPC7987-1G
APOSPC7987-5G
2,2,3,3,4,4,4-Heptafluorobutyltrifluoromethanesulphonate 97%
2,2,3,3,4,4,4-Heptafluorobutyltrifluoromethanesulphonate
Formula:
C₅H₂F₁₀O₃S Boiling Pt: 118…120 °C/732mmHg Storage Temperature: Ambient |
MDL Number:
MFCD02093339 CAS Number: 6401-01-0 UN: 2927 ADR: 6.1+8,II |