4-Fluoro-2-methylbenzenethiol
Supplier: Apollo Scientific
Warning
Synonyms: 5-Fluoro-2-thiotoluene
PC7992-1G PC7992-500MG PC7992-5G
APOSPC7992-1GEA 277 EUR
APOSPC7992-1G APOSPC7992-500MG APOSPC7992-5G
4-Fluoro-2-methylbenzenethiol
4-Fluoro-2-methylthiophenol
Formula: C₇H₇FS
MW: 142.19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06201800
CAS Number: 845823-04-3