Yttrium(III) hexafluoro-2,4-pentanedionate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Yttrium(III) hexafluoroacetylacetonate, Yttrium hexafluoroacetylacetonate
PC8026-5G PC8026-1G PC8026-25G
APOSPC8026-5GEA 174 EUR
APOSPC8026-5G APOSPC8026-1G APOSPC8026-25G
Yttrium(III) hexafluoro-2,4-pentanedionate 98%
Yttrium(III) hexafluoro-2,4-pentanedionate
Formula: C₁₅H₃F₁₈O₆Y
MW: 710.10 g/mol
Melting Pt: 166…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00064764
CAS Number: 18911-76-7