Methyl 5-bromo-4-chloro-2-fluorobenzoate
Supplier: Apollo Scientific
Synonyms:
Methyl 5-bromo-4-chloro-2-fluorobenzoate
PC8038-1G
PC8038-5G
APOSPC8038-1GEA
132
EUR
APOSPC8038-1G
APOSPC8038-5G
Methyl 5-bromo-4-chloro-2-fluorobenzoate
Methyl 5-bromo-4-chloro-2-fluorobenzoate
Formula:
C₈H₅BrClFO₂ MW: 267.4795 g/mol Melting Pt: 75…79 °C Storage Temperature: Ambient |
MDL Number:
MFCD09800902 CAS Number: 951884-02-9 |