4'-(Trifluoromethylthio)acetophenone 98%
Supplier: Apollo Scientific
Warning
PC8042-1G PC8042-5G
APOSPC8042-1GEA 60.9 EUR
APOSPC8042-1G APOSPC8042-5G
4'-(Trifluoromethylthio)acetophenone 98%
4'-(Trifluoromethylthio)acetophenone
Formula: C₉H₇F₃OS
MW: 220.21 g/mol
Melting Pt: 28…31 °C
Storage Temperature: Ambient
MDL Number: MFCD00236332
CAS Number: 713-67-7