2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline
Supplier: Apollo Scientific
Warning
Synonyms: 4-Aminotetrafluorobenzotrifluoride
PC8043-5G PC8043-25G
APOSPC8043-5GEA 234 EUR
APOSPC8043-5G APOSPC8043-25G
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline
Formula: C₇H₂F₇N
MW: 233.09 g/mol
Boiling Pt: 185…187 °C
Density: 1.687 g/cm³ (25 °C)
Flash Pt: 87 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00091518
CAS Number: 651-83-2