4-Bromophenylsulphur pentafluoride
Supplier: Apollo Scientific
Danger
Synonyms: 1-Bromo-4-(pentafluorosulphanyl)benzene
PC8054-1G PC8054-5G
APOSPC8054-1GEA 183 EUR
APOSPC8054-1G APOSPC8054-5G
4-Bromophenylsulphur pentafluoride
4-Bromophenylsulphur pentafluoride
Formula: C₆H₄BrF₅S
MW: 283.06 g/mol
Boiling Pt: 77…78 °C/10mmHg
Flash Pt: 84 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03425928
CAS Number: 774-93-6
UN: 2810
ADR: 6.1 III